Learn More
Cayman Chemical Precelys MInIlys Tubes Bl 1ea
SDP
Catalog No. 502965042
Encompass_Preferred
Package includes everything needed to replace the tubes blocking rod on the Minilys instrument (Item No. 10435). This product is recommended to be replaced once per year or when necessary. [Bertin Catalog S000682-MLYS0-A.0; Previously SP06404-810-NC001.0]
Catalog No. 50-296-5042 Supplier Cayman Chemical Supplier No. 184331 EA
Package includes everything needed to replace the tubes blocking rod on the Minilys instrument (Item No. 10435). This product is recommended to be replaced once per year or when necessary. [Bertin Catalog S000682-MLYS0-A.0; Previously SP06404-810-NC001.0]