Learn More
Cayman Chemical Precelys ZIrconIM OxID Be 1BOX
SDP
Supplier:  Cayman Chemical 139341 BOX
Encompass_Preferred
Includes 100 preps of bulk 0.1 mm ceramic beads. Place in your tubes to optimize your lysing efficiency. Precellys Tissue Homogenizer. Beads are designed for micro-organism lysing like human cells, gram+, bacteria, and more. [Bertin Catalog P000932-LYSK0-A.0; Previously KT03961-1-108.BK]
Catalog No. 50-295-4242