Learn More
Cayman Chemical Precelys ZIrconIM OxID Be 1BOX
SDP
Supplier:  Cayman Chemical 137661 BOX
Encompass_Preferred
Includes 100 preps of bulk 6.8 mm ceramic beads. Place in your tubes to optimize your grinding efficiency. [Bertin Catalog P000931-LYSK0-A.0; Previously KT03961-1-107.BK]
Catalog No. 50-295-5162