Learn More
Cayman Chemical Precelys ZIrconIM OxID Be 1BOX
SDP
Catalog No. 502955437
Encompass_Preferred
Includes 100 preps of bulk 5.0 mm ceramic beads. Place in your tubes to optimize your grinding efficiency. [Bertin Catalog P000930-LYSK0-A.0; Previously KT03961-1-106.BK]
Catalog No. 50-295-5437 Supplier Cayman Chemical Supplier No. 137651 BOX
Includes 100 preps of bulk 5.0 mm ceramic beads. Place in your tubes to optimize your grinding efficiency. [Bertin Catalog P000930-LYSK0-A.0; Previously KT03961-1-106.BK]