Learn More
Sigma Aldrich Fine Chemicals Biosciences Preciflow peristaltic pump1EA
Catalog No. 502909098
Encompass
Traditional peristaltic pumps commonly cause processes to be interrupted because of decreasing flow rates over time aspiration or rupture of tubing by the mechanics causing time and money to be lost. The Preciflow peristaltic pump was developed as a result of twenty years of laboratory experience and involved the systematic elimination of the imperfections found in other pumps on the market. The result is a practical precise economical and reliable pump that is the most compact in its class. Supplied with 5m of silicone tubing 3 / 5mmReplacement of rollers with bearings reduces friction mechanical strain leading to better pump performance and greatly extended tubing lifeAsymmetric pump head reduces pulsationsStepping motor with microprocessor ensures high precision of flow rate with no inertia while turning pump on and offSmall foot print saves bench spaceAll LAMBDA peristaltic pumps have the same following characteristicsDigital speed control in the range
Catalog No. 50-290-9098 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6784061EA
Traditional peristaltic pumps commonly cause processes to be interrupted because of decreasing flow rates over time aspiration or rupture of tubing by the mechanics causing time and money to be lost. The Preciflow peristaltic pump was developed as a result of twenty years of laboratory experience and involved the systematic elimination of the imperfections found in other pumps on the market. The result is a practical precise economical and reliable pump that is the most compact in its class. Supplied with 5m of silicone tubing 3 / 5mmReplacement of rollers with bearings reduces friction mechanical strain leading to better pump performance and greatly extended tubing lifeAsymmetric pump head reduces pulsationsStepping motor with microprocessor ensures high precision of flow rate with no inertia while turning pump on and offSmall foot print saves bench spaceAll LAMBDA peristaltic pumps have the same following characteristicsDigital speed control in the range