Learn More
RWD LIFE SCIENCE INC PRECISE IMPACTOR
SDP
Catalog No. NC3404315
Encompass_Preferred
68099Ⅱ, the new upgraded precise impactor is a very friendly tool for evaluate traumatic brain injury mechanisms in basic medical research. This system is a popular way to make a traumatic brain injury modeling, which is controlled by pneumatic and electric device, and increased accuracy and reproducibility in brain injury modeling. In addition, a unique sensor is used to automatically detect the zero-interface reference point at the impact point of the model accurately and reliably before the start of the impact procedure, thus reducing the tedious process of manual zeroing. In the experiment, the standard cylindrical head hammers with different size are available. The parameters such as impact depth, velocity and dwell time could be precisely controlled to ensure that the damage level to be accurate. It is an accurate tool with high efficiency, repeatability and stability, ensuring the reproducibility of the damage model
Catalog No. NC3404315 Supplier RWD LIFE SCIENCE INC Supplier No. 6809 II
Add to Cart
Add to Cart
Request a Quote
68099Ⅱ, the new upgraded precise impactor is a very friendly tool for evaluate traumatic brain injury mechanisms in basic medical research. This system is a popular way to make a traumatic brain injury modeling, which is controlled by pneumatic and electric device, and increased accuracy and reproducibility in brain injury modeling. In addition, a unique sensor is used to automatically detect the zero-interface reference point at the impact point of the model accurately and reliably before the start of the impact procedure, thus reducing the tedious process of manual zeroing. In the experiment, the standard cylindrical head hammers with different size are available. The parameters such as impact depth, velocity and dwell time could be precisely controlled to ensure that the damage level to be accurate. It is an accurate tool with high efficiency, repeatability and stability, ensuring the reproducibility of the damage model