Learn More
MSC Precision Brand Feeler Gage, 0.0010 inch, Flat, High-Carbon Steel, 25 inch
Catalog No. 5000531796
Encompass
This feeler gage is crafted from high-carbon steel and features a flat leaf design. It has a thickness of 0.0010 inch (0.05 mm) and a length of 25 inch (635 mm), making it suitable for precise measurements in various applications.
  • High-carbon steel material ensures durability and accuracy.
  • Flat leaf style for precise measurements.
  • Precise thickness of 0.0010 inch (0.05 mm) for fine clearance checking.
  • Length of 25 inch (635 mm) provides extended reach.
  • Manufactured in the United States.
Catalog No. 50-005-31796 Supplier MSC Supplier No. 57245532
This feeler gage is crafted from high-carbon steel and features a flat leaf design. It has a thickness of 0.0010 inch (0.05 mm) and a length of 25 inch (635 mm), making it suitable for precise measurements in various applications.
  • High-carbon steel material ensures durability and accuracy.
  • Flat leaf style for precise measurements.
  • Precise thickness of 0.0010 inch (0.05 mm) for fine clearance checking.
  • Length of 25 inch (635 mm) provides extended reach.
  • Manufactured in the United States.