Learn More
MSC Precision Brand Feeler Gage, 0.002 in, 0.5 in, 12 in, High-Carbon Steel
Catalog No. 5000537327
Encompass
This feeler gage features a precise thickness of 0.0020 in and a flat leaf style. Constructed from durable high-carbon steel, it is 0.5 in wide and 12 in long. This package includes 12 individual leaves.
  • Precise 0.002 in thickness
  • Flat leaf style for accurate measurements
  • Durable high-carbon steel construction
  • Measures 0.5 in wide and 12 in long
  • Includes 12 individual leaves
Catalog No. 50-005-37327 Supplier MSC Supplier No. 79649869
This feeler gage features a precise thickness of 0.0020 in and a flat leaf style. Constructed from durable high-carbon steel, it is 0.5 in wide and 12 in long. This package includes 12 individual leaves.
  • Precise 0.002 in thickness
  • Flat leaf style for accurate measurements
  • Durable high-carbon steel construction
  • Measures 0.5 in wide and 12 in long
  • Includes 12 individual leaves