Learn More
MSC Precision Brand Feeler Gages, 0.0025 Inch Thick, 0.5 Inch Wide, 12 Inch Long, High-Carbon Steel, 12 Pack
Catalog No. 5000531499
Encompass
This set of Precision Brand feeler gages is designed with a thickness of 0.0025 decimal inches. Each leaf features a flat style, a length of 12 inches, and a width of 0.5000 decimal inches. The gages are constructed from high-carbon steel, ensuring durability and accuracy.
  • Thickness of 0.0025 "
  • Flat leaf style.
  • Leaf length of 12 inches.
  • Leaf width of 0.5000 "
  • Constructed from high-carbon steel.
  • Set includes 12 leaves.
Catalog No. 50-005-31499 Supplier MSC Supplier No. 79650149
This set of Precision Brand feeler gages is designed with a thickness of 0.0025 decimal inches. Each leaf features a flat style, a length of 12 inches, and a width of 0.5000 decimal inches. The gages are constructed from high-carbon steel, ensuring durability and accuracy.
  • Thickness of 0.0025 "
  • Flat leaf style.
  • Leaf length of 12 inches.
  • Leaf width of 0.5000 "
  • Constructed from high-carbon steel.
  • Set includes 12 leaves.