Learn More
MSC Precision Brand Feeler Gages, High-Carbon Steel, 0.0050 in, 10 Leaves
Catalog No. 5000510818
Encompass
This set of feeler gages provides precise measurements for various applications. It features a specific thickness for accurate gap assessment, durable construction, and a convenient number of leaves for comprehensive use.
  • Provides a thickness of 0.0050 in (0.13 mm)
  • Features a flat leaf style
  • Each leaf measures 5.0000 in (127.00 mm) in length
  • Each leaf measures 0.5 in (12.7 mm) in width
  • Constructed from high-carbon steel for durability
  • Includes 10 individual leaves
Catalog No. 50-005-10818 Supplier MSC Supplier No. 80072408
This set of feeler gages provides precise measurements for various applications. It features a specific thickness for accurate gap assessment, durable construction, and a convenient number of leaves for comprehensive use.
  • Provides a thickness of 0.0050 in (0.13 mm)
  • Features a flat leaf style
  • Each leaf measures 5.0000 in (127.00 mm) in length
  • Each leaf measures 0.5 in (12.7 mm) in width
  • Constructed from high-carbon steel for durability
  • Includes 10 individual leaves