Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Precision™ Circulating Water Baths
SCI_ED

Catalog No. S37372
Change view
Click to view available options
Capacity (Metric):
19 L
35 L
89 L
Model:
Precision CIR 19
Precision CIR 35
Precision CIR 89
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Model
S37372 19 L Precision CIR 19
S37373 35 L Precision CIR 35
S37374 89 L Precision CIR 89
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S37372 Supplier Thermo Scientific™ Supplier No. TSCIR19/EMD
This item is not returnable. View return policy

Achieve outstanding temperature uniformity for your samples with perimeter-directed water flow on Precision circulating water baths. Featuring advanced temperature controller.

Thermo Scientific™ Precision™ circulating water baths are ideal when temperature uniformity and control are particularly critical, such as when working with enzymes and in serological applications. Available in three different models, these high performance baths feature a microprocessor control and include a removable stainless steel gable cover, diffuser tray, and rubber duck.

  • Achieve enhanced temperature uniformity with perimeter-directed water flow
  • Easily clean and maintain bath with coil-free internal design
  • Optimize scheduling with auto-on and auto-off timers
  • Accommodate taller labware with new hinged lid and extended height
  • Help prevent bath overheating and damage with low-fluid protection
  • Easily operate and monitor with icon-based graphical display
  • Protect your work with audible alarms
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 5.0 gal.
Controller Type Digital
Capacity (Metric) 19 L
Certifications/Compliance UL, CE
Temperature Range Ambient +5° to 100°C
Display Monochrome LCD
Heating Capacity 1200 w
Includes 2 Stainless Steel Gable Covers, 2 Diffuser Trays, and Rubber Duck
Material 304 Stainless Steel
Model Precision CIR 19
Temperature Presets 4
Temperature Stability ±0.1°C
Temperature Uniformity ±0.05°C
Dimensions (D x W x H) Interior 12 x 15.3 x 7.6 in.(305 x 387 x 192 mm)
Dimensions (L x W x H) 15.5 x 24.9 x 9.8 in. (394 x 632 x 249 mm)
Electrical Requirements Global Voltage 100/115 V 200/230 V 50/60 Hz
Amperage 9.0-10.5 / 4.3-5.0 (120V/230V)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.