Learn More
Grainger CRC Aviation XT-2000 Precision Cleaner, Aerosol, 12 oz.
Catalog No. 502779672
Encompass_Preferred
This precision cleaner is a high-performance, non-flammable solvent designed for effective maintenance of sensitive electronic equipment. Formulated to safely and quickly remove light oils, dirt, dust, fingerprints, flux, and polar soils, it is ideal for use on contacts, switches, motors, relays, generators, and sensors.
  • Non-flammable aerosol formula for safe application around electrical equipment.
  • Fast evaporation rate to minimize downtime during maintenance.
  • Meets performance specifications of Boeing BSS 7432, Pratt & Whitney SPOP 6, and Douglas CSD #1.
  • Safe for use on most plastics (test on a small area before full application).
  • Provides high dielectric strength of 27,780 Volts.
  • Low odor formulation for improved user comfort during operation.
Catalog No. 50-277-9672 Supplier Grainger Supplier No. 19MY05
This precision cleaner is a high-performance, non-flammable solvent designed for effective maintenance of sensitive electronic equipment. Formulated to safely and quickly remove light oils, dirt, dust, fingerprints, flux, and polar soils, it is ideal for use on contacts, switches, motors, relays, generators, and sensors.
  • Non-flammable aerosol formula for safe application around electrical equipment.
  • Fast evaporation rate to minimize downtime during maintenance.
  • Meets performance specifications of Boeing BSS 7432, Pratt & Whitney SPOP 6, and Douglas CSD #1.
  • Safe for use on most plastics (test on a small area before full application).
  • Provides high dielectric strength of 27,780 Volts.
  • Low odor formulation for improved user comfort during operation.