Learn More
MSC Sprayway Precision Contact Cleaner, Solvent-Based, Safe for Plastics, 20 oz
Catalog No. 502868779
Encompass
This precision cleaning product prevents corrosion by displacing moisture, helping to prevent electrical malfunctions caused by water penetration, humidity, and condensation. It is ideal for applications where lower flash point and higher evaporating solvents may be used.
  • Non-staining
  • Non-corrosive
  • Safe for food packaging and processing equipment
  • Quick drying
  • Leaves no residue
  • Safe on most plastics
  • Effectively removes soil and other contaminants from electronic components
  • VOC compliant CARB/OTC
  • Recyclable packaging
  • Extremely flammable
  • Solvent-based chemistry
  • Colorless
  • Solvent fragrance
Catalog No. 50-286-8779 Supplier MSC Supplier No. 30626345
This precision cleaning product prevents corrosion by displacing moisture, helping to prevent electrical malfunctions caused by water penetration, humidity, and condensation. It is ideal for applications where lower flash point and higher evaporating solvents may be used.
  • Non-staining
  • Non-corrosive
  • Safe for food packaging and processing equipment
  • Quick drying
  • Leaves no residue
  • Safe on most plastics
  • Effectively removes soil and other contaminants from electronic components
  • VOC compliant CARB/OTC
  • Recyclable packaging
  • Extremely flammable
  • Solvent-based chemistry
  • Colorless
  • Solvent fragrance