Learn More
THORLABS INC Precision Cover Glasses, #1.5H Thickness, 22 x 22 mm, Pack of 1000
Catalog No. NC1415511
Encompass
Ground to a uniform thickness of 170 ± 5 µm and are made from Schott D 263 M Glass, an optically clear and colorless glass that exhibits low auto-fluorescence. The uniform thickness and consistent optical properties help reduce image defects due to spherical aberrations. As such, these cover slips are ideal for use with high NA objective lenses.
Catalog No. NC1415511 Supplier THORLABS INC Supplier No. CG15CH
Ground to a uniform thickness of 170 ± 5 µm and are made from Schott D 263 M Glass, an optically clear and colorless glass that exhibits low auto-fluorescence. The uniform thickness and consistent optical properties help reduce image defects due to spherical aberrations. As such, these cover slips are ideal for use with high NA objective lenses.