Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDC Healthcare Blood Recipient System Accessories

Catalog No. 22138867
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-138-867 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-138-867 Supplier PDC Healthcare Supplier No. 220000PDJ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Material Poly
Color White
Product Type Blood Recipient Form
For Use With (Application) Links the patient, the transfused Blood and all the related paperwork to assure safe and complete Blood transfusion
For Use With (Equipment) Securlink™ Blood recipient system
Size Adult/pediatric
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.