Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDC Healthcare Sentry™ LabelBand™ Bar Coded Wristbands

Catalog No. 22275432
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-275-432 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-275-432 Supplier PDC Healthcare Supplier No. 509011PDM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Type Sentry Labelband Wristband
Color White
Includes Tamper evident slit, clear adhesive shield, Media supports text, linear and 2D bar codes, photos and graphics
For Use With (Application) Provides optimal protection from solvents for accurate bar code scanning
Height (English) 1.25 in.
Length (English) 10.75 in.
Length (Metric) 27.3 cm
Closure Type Snap
Closure Material Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.