Learn More
MSC Simple Green Aircraft and Precision Cleaner, Water-Based, Non-Corrosive, 32 oz
Catalog No. 502868303
Encompass
Extreme Aircraft and Precision Cleaner is a non-caustic, water-based solution for high-precision cleaning and degreasing. It features high soil capacity, superior grease-cutting, and a clean-rinsing formula to prevent redeposition and minimize oxidation.
  • Cuts through tough, built-up grease, oil, and dirt without corrosive chemicals
  • Optimized to clean metal, plastic, rubber, and painted surfaces
  • Non-flammable, petroleum-distillate-free, and readily biodegradable formula
  • High soil capacity, superior grease-cutting ability, and clean rinsing formula
  • Safe for structural metals, plastics, paints, or coatings
  • Compatible with oil/water separators and clarifiers
  • VOC compliant formula, contains no Clean Air Act Hazardous Air Pollutants (HAPs)
  • Can be used manually with spray and wipe or mop and bucket
Catalog No. 50-286-8303 Supplier MSC Supplier No. 73239030
Extreme Aircraft and Precision Cleaner is a non-caustic, water-based solution for high-precision cleaning and degreasing. It features high soil capacity, superior grease-cutting, and a clean-rinsing formula to prevent redeposition and minimize oxidation.
  • Cuts through tough, built-up grease, oil, and dirt without corrosive chemicals
  • Optimized to clean metal, plastic, rubber, and painted surfaces
  • Non-flammable, petroleum-distillate-free, and readily biodegradable formula
  • High soil capacity, superior grease-cutting ability, and clean rinsing formula
  • Safe for structural metals, plastics, paints, or coatings
  • Compatible with oil/water separators and clarifiers
  • VOC compliant formula, contains no Clean Air Act Hazardous Air Pollutants (HAPs)
  • Can be used manually with spray and wipe or mop and bucket