Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Precision™ General-Purpose Water Baths

Catalog No. p-4527586
Change view
Click to view available options
Controller Type:
Analog
Model:
184
Capacity (Metric):
19.5 L
Amperage:
5.2 A
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Controller Type Model Capacity (Metric) Amperage
15-474-18 Analog 184 19.5 L 5.2 A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-474-18 Supplier Thermo Scientific™ Supplier No. 2839

Precision General-Purpose Water Baths consist of eight models, including dual chamber and shallow form, with analog or digital control.

Thermo Scientific™ Precision General-Purpose Water Baths consist of eight models, including dual chamber and shallow form, with analog or digital control. Easy-to-maintain, seamless-stainless steel interior chamber and epoxy powder-coated exterior are resistant to corrosion and chemical damage.

Rugged, high-performance baths maintain water temperature from ambient to 95°C or 99.9°C (depending on model) with ±0.2°C uniformity (at 37°C) and ±0.1°C control resolution (at 37°C) with stainless-steel gable cover.
  • Choice of analog or digital temperature control
  • Chamber capacities ranging from 1.5 to 43L
  • Front-mounted controls simplify operation
  • Overtemperature safety circuitry prevents thermal runaway
  • Dual-chamber models have independent controls for operating at different temperatures

Analog Baths

  • Easy-to-use analog controls and hydraulic thermostat are ideal for applications in which temperature setpoints seldom change
  • Backup high-limit safety thermostat
  • Illuminated power switch

Digital Baths

  • Allow temperature setpoint with ±0.1°C precision at the push of a button
  • Microprocessor control with bright three-digit LED temperature readout
  • Automatic overtemperature protection

Includes:

Gable cover: stainless steel for all except Models 181 and 281, which are polypropylene; Diffuser shelf; Spirit-filled thermometer (analog baths only); Rubber duck

Recommended for:

  • Bacteriological examinations
  • Coagulation tests
  • Copper strip corrosion tests
  • Crude oil studies
  • Cytochemistry
  • Demulsibility studies
  • Environmental studies
  • Food processing QC
  • Genetic studies
  • Hormone studies
  • Immunological research
  • Incubation for microbiological assays
  • Incubation for microcentrifugation tubes
  • Melting agar
  • Metallurgical analysis
  • Molecular biology
  • Protein analysis
  • Radioactive isotope uptake studies
  • Radiochemistry
  • Serological research
  • Thawing
  • Thawing blood
  • Thawing cryopreservation vials
  • Tissue culture research
  • Virology research
  • Warming reagents
  • Water quality research
TRUSTED_SUSTAINABILITY

Specifications

Chamber Material Stainless Steel
Controller Type Analog
Capacity (Metric) 19.5 L
Certifications/Compliance UL listed
Temperature Range Ambient to 95° or 99°C
Material Epoxy Powder-coated Steel
Model 184
Temperature Resolution ±0.1‡°C
Temperature Uniformity ±0.2°C
Dimensions (L x W x H) 19.7 x 15.75 x 9.75 in. (49.9 x 40 x 24.8 cm)
Length (English) 19.7 in.
Length (English) Interior 14 in.
Length (Metric) 49.9 cm
Length (Metric) Interior 35.6 cm
Width (English) 15.75 in.
Width (English) Interior 12 in.
Width (Metric) 40 cm
Width (Metric) Interior 30.5 cm
Height (English) 9.75 in.
Height (Metric) 24.8 cm
Depth (English) Interior 8 in.
Depth (Metric) Interior 20.3 cm
Shipping Weight (English) 25 lb.
Shipping Weight (Metric) 11.3 kg
Voltage 120 V
Wattage 600 w
Amperage 5.2 A
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.