Learn More
ABBOTT DIABETES CARE INC PRECISION GLUCOSE STRIPS 50PK
Catalog No. NC1921582
Encompass
PRECISION XTRA BLOOD GLUCOSE TEST STRIPS. 50 count. Sample size: 0.6 µLResults in 5 secondsNo coding requiredIndividually wrapped test strips for easy testing on-the-goPrecision Xtra meter sold separately
This item is not returnable. View return policy
Catalog No. NC1921582 Supplier ABBOTT DIABETES CARE INC Supplier No. 9972865
PRECISION XTRA BLOOD GLUCOSE TEST STRIPS. 50 count. Sample size: 0.6 µLResults in 5 secondsNo coding requiredIndividually wrapped test strips for easy testing on-the-goPrecision Xtra meter sold separately