Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Precision LentiORF NFE2L2 w/o Stop Codon, Unit Size: 2 x 25 µl viral particles
Catalog No. NC3735628
Encompass
This gene encodes a transcription factor which is a member of a small family of basic leucine zipper (bZIP) proteins. The encoded transcription factor regulates genes which contain antioxidant response elements (ARE) in their promoters; many of these genes encode proteins involved in response to injury and inflammation which includes the production of free radicals. Multiple transcript variants encoding different
Catalog No. NC3735628 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. OHS5900202624558
This gene encodes a transcription factor which is a member of a small family of basic leucine zipper (bZIP) proteins. The encoded transcription factor regulates genes which contain antioxidant response elements (ARE) in their promoters; many of these genes encode proteins involved in response to injury and inflammation which includes the production of free radicals. Multiple transcript variants encoding different