Learn More
Electron Microscopy Sciences Tripod Polisher PrecISIon Locking Micrometer
SDP
Catalog No. 50980865
Encompass
Special larger diameter locking rings for easy locking and are graduated in one micron increments with 13mm travel. Non-coating Micrometers not recommended. - For use with Tripod Polisher.
Catalog No. 50-980-865 Supplier Electron Microscopy Sciences Supplier No. 5011610
Special larger diameter locking rings for easy locking and are graduated in one micron increments with 13mm travel. Non-coating Micrometers not recommended. - For use with Tripod Polisher.