Learn More
Cole-Parmer Traceable Precision Monitoring Thermohygrometer with Calibration
Catalog No. NC1902681 Shop All Cole-Parmer Products
Encompass_Preferred
Simultaneously display Min/Max and current temperature and humidity with 7-day graph. Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory. Scroll through temperature and humidity data for the past seven days. Maintain data integrity-historical data cannot be cleared, only overwritten. Easily switch between F/C and 12/24-hour time formats.
Catalog No. NC1902681 Supplier Cole-Parmer Supplier No. 1800434
Simultaneously display Min/Max and current temperature and humidity with 7-day graph. Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory. Scroll through temperature and humidity data for the past seven days. Maintain data integrity-historical data cannot be cleared, only overwritten. Easily switch between F/C and 12/24-hour time formats.