Learn More
Grainger Starrett 436.1 Outside Micrometer, 17 in. to 18 in. Range, 0.001 in. Resolution, 1 ea.
Catalog No. 502786422
Encompass_Preferred
The outside micrometer is a precision measurement tool designed for reliable, accurate performance in industrial and workshop environments. It features a robust, balanced frame and thimble for optimal handling and consistent results.
  • Carbide-tipped flat anvil and spindle
  • Advanced sleeve design with staggered lines and distinct figures for precise, easy readability
  • Ratchet thimble for consistent pressure
  • Knurled ring-type lock nut for secure adjustments
  • Accuracy of ±.0003 in.
  • Resolution of .001 in.
  • Range of 17 in. to 18 in.
  • Includes storage case
Catalog No. 50-278-6422 Supplier Grainger Supplier No. 197M48
The outside micrometer is a precision measurement tool designed for reliable, accurate performance in industrial and workshop environments. It features a robust, balanced frame and thimble for optimal handling and consistent results.
  • Carbide-tipped flat anvil and spindle
  • Advanced sleeve design with staggered lines and distinct figures for precise, easy readability
  • Ratchet thimble for consistent pressure
  • Knurled ring-type lock nut for secure adjustments
  • Accuracy of ±.0003 in.
  • Resolution of .001 in.
  • Range of 17 in. to 18 in.
  • Includes storage case