Learn More
Grainger Precision Pentalobe Screwdriver PL4
Catalog No. 502764503
Encompass_Preferred
The WIHA Precision Pentalobe Screwdriver is a high quality screwdriver designed for use with Apple devices that have Pentalobe screws. It features a black chrome-plated shaft with a precision machined tip that provides exact fit and reduces cam-out. The screwdriver has a soft grip handle for comfort and control and comes with a protective cap. It is made in Germany and measures 4 inches long with a tip size of 0.8mm. This professional grade screwdriver allows for easy repair and disassembly of MacBooks, iPhones, iPads and other Apple products.
Catalog No. 50-276-4503 Supplier Grainger Supplier No. 16NG83
The WIHA Precision Pentalobe Screwdriver is a high quality screwdriver designed for use with Apple devices that have Pentalobe screws. It features a black chrome-plated shaft with a precision machined tip that provides exact fit and reduces cam-out. The screwdriver has a soft grip handle for comfort and control and comes with a protective cap. It is made in Germany and measures 4 inches long with a tip size of 0.8mm. This professional grade screwdriver allows for easy repair and disassembly of MacBooks, iPhones, iPads and other Apple products.