Learn More
Grainger Wiha Pentalobe Precision Screwdriver, PL2 Tip, 40mm Blade, 1 ea
Catalog No. 502789229
Encompass_Preferred
This precision screwdriver features a PL2 pentalobe tip designed for high-performance applications. Constructed from high-performance tool steel with a hard-chromed finish on the blades and tips, it offers exceptional durability and wear resistance. The tool is engineered for accuracy and comfort during delicate assembly or repair tasks.
  • Precision machined pentalobe tips for an exact fit
  • High-performance tool steel construction
  • Hard-chromed finish on blades and tips
  • Tapered ergonomic handle for improved control
  • Easy spin finger tip control cap
  • Designed to fit fasteners used in Apple brand products
Catalog No. 50-278-9229 Supplier Grainger Supplier No. 16ML73
This precision screwdriver features a PL2 pentalobe tip designed for high-performance applications. Constructed from high-performance tool steel with a hard-chromed finish on the blades and tips, it offers exceptional durability and wear resistance. The tool is engineered for accuracy and comfort during delicate assembly or repair tasks.
  • Precision machined pentalobe tips for an exact fit
  • High-performance tool steel construction
  • Hard-chromed finish on blades and tips
  • Tapered ergonomic handle for improved control
  • Easy spin finger tip control cap
  • Designed to fit fasteners used in Apple brand products