Learn More
Grainger Precision Pentalobe Screwdriver PL5
Catalog No. 502763728
Encompass_Preferred
The WIHA Precision Pentalobe Screwdriver is a high quality screwdriver designed for use with Apple products that feature pentalobe screws. It has a precision machined tip that provides exact fit and avoids cam-out. The screwdriver features a chrome-plated shaft and an ergonomic, soft grip handle. It comes in a compact size of only 3-3/4 inches long. This professional quality screwdriver allows for easy repair and disassembly of Apple devices.
Catalog No. 50-276-3728 Supplier Grainger Supplier No. 16NG84
Add to Cart
Add to Cart
The WIHA Precision Pentalobe Screwdriver is a high quality screwdriver designed for use with Apple products that feature pentalobe screws. It has a precision machined tip that provides exact fit and avoids cam-out. The screwdriver features a chrome-plated shaft and an ergonomic, soft grip handle. It comes in a compact size of only 3-3/4 inches long. This professional quality screwdriver allows for easy repair and disassembly of Apple devices.