Learn More
Azzota Corp Precision pH meter ORP -2-20
Catalog No. 5000887291
Encompass_Preferred
Benchtop Precision pH/mV/C/ F Meter pH Range -2 00-20 00pH mV Range /-1999 9mV with ORP electrode
Catalog No. 50-008-87291 Supplier Azzota Corp Supplier No. PHMV920ORP
Benchtop Precision pH/mV/C/ F Meter pH Range -2 00-20 00pH mV Range /-1999 9mV with ORP electrode