Learn More
Electron Microscopy Sciences PrecISIon Pin Vise, 4/Set
SDP
Catalog No. 50996502
Encompass_Preferred
These tool steel collets grip firmly when screw chucks are tightened. The chromium-plated handle is hollow which allows for the holding of long wires. The body is stepped, with the small portion knurled allowing for rotation between the fingers. The larger portion is smooth and it easily can be held firmly in lathes and drill presses. 4 sizes of chuck openings.
Catalog No. 50-996-502 Supplier Electron Microscopy Sciences Supplier No. 6207410
These tool steel collets grip firmly when screw chucks are tightened. The chromium-plated handle is hollow which allows for the holding of long wires. The body is stepped, with the small portion knurled allowing for rotation between the fingers. The larger portion is smooth and it easily can be held firmly in lathes and drill presses. 4 sizes of chuck openings.