Learn More
Bio Rad Laboratories Precision Plus Protein Unstained Protein Standards, Strep-tagged, recombinant, 5 mL
Catalog No. NC1203903
Encompass
5 ml (5 x 1 ml), unstained mixture of ten Strep-tagged, recombinant proteins (10–250 kD), including three reference bands (25, 50, 75 kD), 500 applications.
Catalog No. NC1203903 Supplier Bio Rad Laboratories Supplier No. 1610396
5 ml (5 x 1 ml), unstained mixture of ten Strep-tagged, recombinant proteins (10–250 kD), including three reference bands (25, 50, 75 kD), 500 applications.