Learn More
Bio Rad Laboratories PRECISION PLUS PROTEIN WESTERN
Catalog No. 509010162
Encompass
Mini-PROTEAN TGX extended shelf-life gels are precast polyacrylamide gels for vertical SDS-PAGE and native PAGE protein separations. Gels 8.6 cm x 7.2 cm x 1.0 mm (W x L x thickness) are supplied in a 10.0 x 8.5 cm cassette.
This item is not returnable. View return policy
Catalog No. 50-901-0162 Supplier Bio Rad Laboratories Supplier No. 1610376
Mini-PROTEAN TGX extended shelf-life gels are precast polyacrylamide gels for vertical SDS-PAGE and native PAGE protein separations. Gels 8.6 cm x 7.2 cm x 1.0 mm (W x L x thickness) are supplied in a 10.0 x 8.5 cm cassette.