Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Precision and Proof : Forensic Biotechnology Skills Training
SDPSCI_ED

Catalog No. S001097 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
9+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S001097 9+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001097 Supplier EDVOTEK™ Supplier No. 197
Add to Cart
Add to Cart

Step into the role of a forensic investigator with this hands-on forensic biotechnology skills training kit, students will explore key techniques used to analyze physical evidence, from fingerprint analysis to blood spatter patterns.

  • Practice real forensic methods used in crime scene investigations.
  • Use micropipettes in a Pipetting-by-Numbers™ activity to compare fingerprints.
  • Reveal the correct fingerprint match under UV or blue light.
  • Recreate and study blood spatter patterns using transfer pipets.
  • Investigate how height and force affect blood droplet shape.
  • Group Size: For 10 Lab Groups.
  • Kit Includes: Instructions, Various Dyes, Pipetting-by-Numbers™ Canvas, Microcentrifuge Tubes, Simulated Blood Spatter Solution, Transfer Pipets.
  • All You Need: Automatic Pipettes, UV or Blue Light, Small Containers for Discarding Used Tips, Scissors, Tape.
  • Storage: Room Temperature.

Specifications

Grade Level Intermediate - Middle School, High School
Grade 9+
Class Size 10 Lab Groups
Product Type Forensic Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.