Learn More
Cole-Parmer Extech RH490 Precision Psychrometer (GPP, g/kg)
Supplier:  Cole-Parmer 3785101
Encompass
Precision capacitance humidity sensor offers fast response. Slim design with rubberized sides for better grip and easy, one-handed operation. The dual LCD provides simultaneous display of humidity and temperature, or humidity and dew point, or humidity and wet bulb. Advanced features include data hold, min/max readings, low-battery indication, and auto power off.
Catalog No. 50-993-044