Learn More
PCE AMERICAS INC Precision balance scale with piece counting function
SDP
Catalog No. NC3654598
Encompass_Preferred
"The counting scale is a scale for daily laboratory use. Another function besides the normal measuring function of the counting scale is the piece counting function. This function allows the counting scale to determine the number of screws, for example. Thus, the counting scale can also be used in logistics, for example. So that the display is readable even in low light conditions, the display of the counting scale has a backlight. In addition to the measuring functions, the counting scale also has an RS 232 interface. Thanks to this interface, the measured values ​​can be transferred to a PC, for example. This counting scale can be used for stationary as well as for mobile use. For mobile use, the counting scale requires 3 x 1.5V AA batteries, which can be used quickly and easily."
Catalog No. NC3654598 Supplier PCE AMERICAS INC Supplier No. PCEBSK 5100
"The counting scale is a scale for daily laboratory use. Another function besides the normal measuring function of the counting scale is the piece counting function. This function allows the counting scale to determine the number of screws, for example. Thus, the counting scale can also be used in logistics, for example. So that the display is readable even in low light conditions, the display of the counting scale has a backlight. In addition to the measuring functions, the counting scale also has an RS 232 interface. Thanks to this interface, the measured values ​​can be transferred to a PC, for example. This counting scale can be used for stationary as well as for mobile use. For mobile use, the counting scale requires 3 x 1.5V AA batteries, which can be used quickly and easily."