Learn More
Scientific Industries PRECISION SCALE 220G
Catalog No. 50728283
Encompass_Preferred
220g x 0.001g, compact precision laboratory scale with small footprint with high precision and accuracy. ABS housing, USB, RS232 interface.
Catalog No. 50-728-283 Supplier Scientific Industries Supplier No. ATA220
220g x 0.001g, compact precision laboratory scale with small footprint with high precision and accuracy. ABS housing, USB, RS232 interface.