Learn More
Sigma Aldrich Fine Chemicals Biosciences Precision SealR rubber sep
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z553999100EA
Encompass
The recommended choice for use with organic chemicals and solvents pressure applications and cannulation techniquesPrecision Seal septa are designed specifically for the rigorous requirements of chemical applications.Precision fit Precision Seal septa have a longer tapered plug that seats firmly inside the glass joint or tube providing a sealing surface that is 50 to 80% greater than standard sleeve stoppers. The inner seal design of Precision Seal septa minimizes chemical and solvent vapor migration up the joint to the sleeve which can swell leak or fail from exposure. The surface texture of the rubber is similar to that of a ground glass joint and resists movement once seated.Non-contaminating Precision Seal natural rubber is lower in solvent extractables 1.9% vs. standard sleeve stoppers 5-6%. Precision Seal rubber is higher in density to minimize the effects of solvent degradation. They are manufactured from a single certified lot for absolute consistency.Safet
Catalog No. 50-290-5660