Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Precision™ Shaking Water Baths
GSA_VASCI_ED

Catalog No. S37368
Change view
Click to view available options
Capacity (Metric):
15 L
27 L
Model:
Precision SWB 15
Precision SWB 27
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Model
S37368 15 L Precision SWB 15
S37369 27 L Precision SWB 27
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S37368 Supplier Thermo Scientific™ Supplier No. TSSWB15/EMD
This item is not returnable. View return policy

Shaking water baths support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culturing and genetics sequencing.

Thermo Scientific™ Precision™ shaking water baths support a range of sensitive life science and QA/QC applications – from warming fragile reagents to tissue culturing and genetics sequencing. Easily clean and maintain your bath with new coil-free interior. The Precision shallow form shaking bath has a removable tray that allows for use with smaller sample containers. Precision Dubnoff shaking water baths are designed specifically for applications that require your samples to be incubated in a controlled atmosphere.

  • Easily clean and maintain bath with coil-free design
  • Optimize scheduling with auto-on and auto-off timers
  • Help prevent bath overheating and damage with low-fluid protection
  • Conveniently save commonly used settings with four temperature and shaking speed presets
  • Help protect your work with audible alarms
  • Easily operate and monitor with icon-based graphical display
  • Accommodate taller labware with new hinged lid and extended height
  • Features next-generation shaker motor

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Chamber Material 304 Stainless Steel
Controller Type Digital
Capacity (Metric) 15 L
Certifications/Compliance UL, CE
Temperature Range Ambient +5° to 100°C
Type Shaking Water Bath
Display Monochrome LCD
Heating Capacity 1200 w
Includes Stainless Steel Gable Cover, Shaking Tray, and Rubber Duck
Material Epoxy Powder-coated Cold Rolled Steel
Model Precision SWB 15
Temperature Presets 4
Temperature Stability ±0.1°C
Temperature Uniformity ±0.05°C
Dimensions (L x W x H) 15.5 x 24.9 x 9.8 in. (394 x 632 x 249 mm)
Electrical Requirements Global Voltage 100/115 V 200/230 V 50/60 Hz
Amperage 9.0-10.5 / 4.3-5.0 (120V/230V)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.