Learn More
Peak Scientific Instruments Ltd Precision SL 200 Hydrogen Generator, Auto Fill, Black Supports GC/FID
SDP
Catalog No. 5000804192
Encompass_Preferred
2 models – 100cc and 200cc
99.9995% purity @ 100 psi
One-button start-up and shutdown
Easy end-user managed servicing
Optional auto-water fill or pressurized water fill
Auto-shutdown fail-safe feature
CE, FCC and CSA compliant
2 year comprehensive warranty
Available in black or white
This item is not returnable. View return policy
Catalog No. 50-008-04192 Supplier Peak Scientific Instruments Ltd Supplier No. 3302953
2 models – 100cc and 200cc
99.9995% purity @ 100 psi
One-button start-up and shutdown
Easy end-user managed servicing
Optional auto-water fill or pressurized water fill
Auto-shutdown fail-safe feature
CE, FCC and CSA compliant
2 year comprehensive warranty
Available in black or white