Learn More
World Precision Instrument Precision Stereo Zoom Binocular Microscope (III) on Boom Stand
SDP
Catalog No. 502538616
Encompass
The PZMIII offers stereo viewing with ample working distance when used with the long working distance (LWD) option. The long working distance (LWD) option with the 501371 and 501375 on a trinocular microscope shows nearly the same scene on the screen as the viewer sees in the eyepieces.
Catalog No. 50-253-8616 Supplier World Precision Instrument Supplier No. PZMIIIBS
The PZMIII offers stereo viewing with ample working distance when used with the long working distance (LWD) option. The long working distance (LWD) option with the 501371 and 501375 on a trinocular microscope shows nearly the same scene on the screen as the viewer sees in the eyepieces.