Learn More
Cole-Parmer Aven Tools Tweezers, stainless steel, straight, ultra fine sharp tips, 4-1/4"L
Catalog No. NC9804649 Shop All Cole-Parmer Products
Encompass_Preferred
Recommended for micro electronics and precision assembly, these finely crafted stainless steel tweezers feature hand finished points for accuracy and excellent tip hardness. Anti-glare finish reduces eye strain. Anti-magnetic, anti-acid, and acid resistant.
Catalog No. NC9804649 Supplier Cole-Parmer Supplier No. 0728706
Recommended for micro electronics and precision assembly, these finely crafted stainless steel tweezers feature hand finished points for accuracy and excellent tip hardness. Anti-glare finish reduces eye strain. Anti-magnetic, anti-acid, and acid resistant.