Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Precision Tweezers With Offset Fine Tips

Catalog No. 21125088 Shop All Surgical Design Products
Change view
Click to view available options
Length (English):
4.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
21-125-088 4.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-125-088 Supplier Surgical Design™ Supplier No. SPL05A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Precision tweezers with offset fine tips, 4.5 in., crafted from antimagnetic 300-series stainless steel for exceptional control in microscopy, histology, and electronics work.

  • Offset fine tips provide enhanced visibility and access when working with small or delicate materials.
  • 4.5 in. compact length ensures stable, precise handling during detailed tasks.
  • Made from antimagnetic 300-series stainless steel for corrosion resistance, durability, and non-reactive performance.
  • Excellent for microscopy, histology, tissue manipulation, electronic assembly, and slide preparation.
  • Smooth, controlled tension for comfortable, fatigue-free use.
  • Reusable, easy-to-clean construction for long-term professional reliability.

Specifications

Product Type General Purpose Tweezers
Material Stainless Steel (304)
Style High Precision Tweezers
Tip Style Offset
Length (English) 4.5 in.
Length (Metric) 11.4 cm
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.