Learn More
Andwin Scientific PRECISION WIPES TISSUE WIPERS
SDP
Catalog No. NC9160131
Encompass_Preferred
The KIMTECH SCIENCE Precision Wipes are gentle, low-linting, economical wipers that are ideal for use in laboratories. It has a 1-ply thickness and a dimension of 11.2 x 21.3 cm (413/32 x 813/32"). There are 280 wipes in one box.
Catalog No. NC9160131 Supplier Andwin Scientific Supplier No. 21903025
The KIMTECH SCIENCE Precision Wipes are gentle, low-linting, economical wipers that are ideal for use in laboratories. It has a 1-ply thickness and a dimension of 11.2 x 21.3 cm (413/32 x 813/32"). There are 280 wipes in one box.