Learn More
McKesson General Medical PRECISION XTRA TEST STRIP 10PK
Catalog No. NC1881358
Encompass
Blood Ketone Test Strips Precision Xtra® 10 Strips per Box 10 second test time and 1.5 microliters blood sample size For Ketone Precision Meters
This item is not returnable. View return policy
Catalog No. NC1881358 Supplier McKesson General Medical Supplier No. 647191
Blood Ketone Test Strips Precision Xtra® 10 Strips per Box 10 second test time and 1.5 microliters blood sample size For Ketone Precision Meters