Learn More
Bound Tree Medical Precision Xtra Test Strips, Capillary Only. Box of 100
Catalog No. NC0767828
Encompass_Preferred
Precision Xtra Test Strips, Capillary, Individually Wrapped, 100/Box. The Precision Xtra blood glucose test strips, designed with TrueMeasure Technology, feature the following benefits: Now no coding required, Small 0.6 microliter sample size, Simple blood application.
Catalog No. NC0767828 Supplier Bound Tree Medical Supplier No. 5289987835
Precision Xtra Test Strips, Capillary, Individually Wrapped, 100/Box. The Precision Xtra blood glucose test strips, designed with TrueMeasure Technology, feature the following benefits: Now no coding required, Small 0.6 microliter sample size, Simple blood application.