Learn More
Peak Scientific Instruments Ltd Precision Zero Air 1.5L Produces up to 1.5L/min of Zero Air
SDP
Catalog No. 5000804184
Encompass_Preferred
Integrates with existing Precision products, with same modular, stackable design
Hydrocarbon content (as Methane) < 0.05ppm for market-leading purity
Up to 1.5L/min of Zero Air
Up to 80psi/5.5bar outlet pressure
Minimum maintenance requirements, no expensive catalyst chamber replacements
Greatly reduced risk of contaminants entering system and no risk of running out of gas
Stylish and contemporary aesthetics and functional design
Status indicative lighting feature (amber warmup, green ready)
Compatible with Precision Air Compressor
12 month comprehensive on-site warranty
Catalog No. 50-008-04184 Supplier Peak Scientific Instruments Ltd Supplier No. 601501
Integrates with existing Precision products, with same modular, stackable design
Hydrocarbon content (as Methane) < 0.05ppm for market-leading purity
Up to 1.5L/min of Zero Air
Up to 80psi/5.5bar outlet pressure
Minimum maintenance requirements, no expensive catalyst chamber replacements
Greatly reduced risk of contaminants entering system and no risk of running out of gas
Stylish and contemporary aesthetics and functional design
Status indicative lighting feature (amber warmup, green ready)
Compatible with Precision Air Compressor
12 month comprehensive on-site warranty