Learn More
Medchemexpress LLC Preclamol hydrochloride ((-)-3-PPP hydrochloride) | 88768-67-6 | 98.0% | 255.78 | 1 ML
SDP
Catalog No. 5000346608
Encompass_Preferred
Preclamol hydrochloride ((-)-3-PPP hydrochloride) is a selective dopamine autoreceptor agonist. It is suitable for research related to schizophrenia.
  • Acts as a selective dopamine autoreceptor agonist.
  • Has potential for schizophrenia research.
Catalog No. 50-003-46608 Supplier Medchemexpress LLC Supplier No. HY108698100MG
Preclamol hydrochloride ((-)-3-PPP hydrochloride) is a selective dopamine autoreceptor agonist. It is suitable for research related to schizophrenia.
  • Acts as a selective dopamine autoreceptor agonist.
  • Has potential for schizophrenia research.