Learn More
Medchemexpress LLC Preclamol hydrochloride | 88768-67-6 | 98.0% | 255.78 | 1 MG
SDP
Catalog No. 5000345124
Encompass_Preferred
Preclamol hydrochloride, also known as (-)-3-PPP hydrochloride, is a selective dopamine autoreceptor agonist used for research purposes. It functions as a selective dopamine autoreceptor agonist and shows potential for research related to schizophrenia.
  • Functions as a selective dopamine autoreceptor agonist
  • Suitable for research use only
Catalog No. 50-003-45124 Supplier Medchemexpress LLC Supplier No. HY10869810MG
Preclamol hydrochloride, also known as (-)-3-PPP hydrochloride, is a selective dopamine autoreceptor agonist used for research purposes. It functions as a selective dopamine autoreceptor agonist and shows potential for research related to schizophrenia.
  • Functions as a selective dopamine autoreceptor agonist
  • Suitable for research use only