Learn More
Medchemexpress LLC Preclamol hydrochloride | 88768-67-6 | 98.0% | 255.78 | 10 MG
SDP
Catalog No. 5000345505
Encompass_Preferred
Preclamol hydrochloride is a selective dopamine autoreceptor agonist, researched for its potential applications in the study of schizophrenia.
  • Functions as a selective dopamine autoreceptor agonist.
  • Has potential for research related to schizophrenia.
Catalog No. 50-003-45505 Supplier Medchemexpress LLC Supplier No. HY10869850MG
Preclamol hydrochloride is a selective dopamine autoreceptor agonist, researched for its potential applications in the study of schizophrenia.
  • Functions as a selective dopamine autoreceptor agonist.
  • Has potential for research related to schizophrenia.