Learn More
Medchemexpress LLC Preclamol hydrochloride | 88768-67-6 | MFCD00069366 | 98.0% | 255.78 g/mol | C14H22ClNO | 5 MG
SDP
Catalog No. 5000263490
Encompass_Preferred
Preclamol hydrochloride is a research compound that functions as a selective dopamine autoreceptor agonist. It is used in preclinical pharmacology to probe dopamine receptor activity and has been investigated for applications related to schizophrenia research. The substance is provided in solid form and as a ready-to-use DMSO solution for laboratory assays.
  • Selective dopamine autoreceptor agonist.
  • Used for preclinical studies of dopamine receptor function.
  • Investigated in schizophrenia-related research.
  • Supplied as a solid and as a 10 mM solution in DMSO.
  • High purity: 98.0%.
  • Molecular formula C14H22ClNO; molecular weight 255.78 g/mol.
  • White to light yellow solid appearance.
Catalog No. 50-002-63490 Supplier Medchemexpress LLC Supplier No. HY1086985MG
Preclamol hydrochloride is a research compound that functions as a selective dopamine autoreceptor agonist. It is used in preclinical pharmacology to probe dopamine receptor activity and has been investigated for applications related to schizophrenia research. The substance is provided in solid form and as a ready-to-use DMSO solution for laboratory assays.
  • Selective dopamine autoreceptor agonist.
  • Used for preclinical studies of dopamine receptor function.
  • Investigated in schizophrenia-related research.
  • Supplied as a solid and as a 10 mM solution in DMSO.
  • High purity: 98.0%.
  • Molecular formula C14H22ClNO; molecular weight 255.78 g/mol.
  • White to light yellow solid appearance.