Learn More
Cole-Parmer Cole-Parmer Precleaned EPA Clear Wide-Mouth Septa Jars, 60 mL, 24/cs
Catalog No. NC1640763 Shop All Cole-Parmer Products
Encompass_Preferred
Glass Septa Jars are processed and certified to meet the requirements of the current US EPA "Specifications and Guidance for Contaminant-Free Sample Containers" document for purgeable volatile organic analysis. Include PTFE-lined silicone septa bonded to open-top caps.
Catalog No. NC1640763 Supplier Cole-Parmer Supplier No. 9954020
Glass Septa Jars are processed and certified to meet the requirements of the current US EPA "Specifications and Guidance for Contaminant-Free Sample Containers" document for purgeable volatile organic analysis. Include PTFE-lined silicone septa bonded to open-top caps.