Learn More
Cole-Parmer Essentials Pre-Cleaned EPA Sample Jar, Clear Glass, 500mL (17 oz); 12/CS
Catalog No. NC1985167 Shop All Cole-Parmer Products
Encompass_Preferred
These pre-cleaned, narrow-mouth jars are processed and certified to meet the requirements of the current US EPA Specifications and Guidance for Contaminant-Free Sample Containers document for semi-volatile organics, pesticides, PCBs, metals, cyanide, fluoride, nitrate, and nitrite analyses.
Catalog No. NC1985167 Supplier Cole-Parmer Supplier No. 9954043
These pre-cleaned, narrow-mouth jars are processed and certified to meet the requirements of the current US EPA Specifications and Guidance for Contaminant-Free Sample Containers document for semi-volatile organics, pesticides, PCBs, metals, cyanide, fluoride, nitrate, and nitrite analyses.